Please login to your online account to display your discounted pricing

CD98 Rat anti-Mouse, PE, Clone: RL388, eBioscience™

Rat Monoclonal Antibody

$158.05 - $448.05

Specifications

Antigen CD98
Clone RL388
Host Species Rat
Gene Alias 4F2, Ly-10
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010531

 
life technologies
LS12098181
50μg Each for $158.05

5010533

 
life technologies
LS12098183
200μg Each for $448.05
Description & Specifications

Specifications

Antigen CD98
Clone RL388
Host Species Rat
Gene Alias 4F2, Ly-10
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The RL388 monoclonal antibody reacts with mouse CD98, also known as 4F2 antigen and Ly-10. RL388 detects a disulfide-linked heterodimer complex (130 kDa) composed of a glycosylated heavy (86 kDa) and a nonglycosylated light (39 kDa) subunits. CD98 is expressed on the surface of activated lymphocytes and is involved in cell activation and calcium flux events.