Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD95/Fas Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD95/Fas Ab-3, Mouse Monoclonal Antibody.

$705.51 - $1,554.08

Specifications

Antigen CD95/Fas Ab-3
Clone GM30
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1098S

 
thermo scientific™ lab vision™
MS1098S
1mL Each for $1,554.08

MS1098S0

 
thermo scientific™ lab vision™
MS1098S0
100µL Each for $705.51

MS1098S1

 
thermo scientific™ lab vision™
MS1098S1
500µL Each for $1,202.55
Description & Specifications

Specifications

Antigen CD95/Fas Ab-3
Clone GM30
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Apoptosis
Type Primary
Immunogen Recombinant protein corresponding to the internal domain near the C-terminal end of the Fas molecule
Isotype IgG1
Purification Method Small intestine

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD95/Fas Ab-3, Mouse Monoclonal Antibody.

CD95, also known as Fas, is a transmembrane glycoprotein. It is a member of the nerve growth factor receptor/tumor necrosis factor superfamily. This cell surface molecule mediates receptor-triggered apoptosis. The Fas antigen is expressed on the surface of various cell types, including activated T and B lymphocytes and T lymphoblastoid cell lines.

Host Species: Mouse

Clone: GM30

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Internal domain near the C-terminus

Immunogen: Recombinant protein corresponding to the internal domain near the C-terminal end of the Fas molecule

Molecular Weight: 48 kDa

Positive Control: Small intestine

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO