Please login to your online account to display your discounted pricing

CD95 (APO-1/Fas) Mouse anti-Mouse, Biotin, Clone: 15A7, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $361.05

Specifications

Antigen CD95 (APO-1/Fas)
Clone 15A7
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011674

 
life technologies
LS13095181
50μg Each for $129.05

5011675

 
life technologies
LS13095185
500μg Each for $361.05
Description & Specifications

Specifications

Antigen CD95 (APO-1/Fas)
Clone 15A7
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 15A7 monoclonal antibody reacts with mouse CD95 (Fas, Apo-1), a 40-50kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, activated lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. 15A7 reacts with mouse CD95 in BALB/c strain and a bit more weakly in C57Bl/6 strain. 15A7 does not induce apoptosis of Fas-expressing cells.