Please login to your online account to display your discounted pricing

CD95 (APO-1/Fas) Mouse anti-Human, PE-Cy5, Clone: DX2, eBioscience™

Mouse Monoclonal Antibody

$158.05 - $375.55

Specifications

Antigen CD95 (APO-1/Fas)
Clone DX2
Host Species Mouse
Species Reactivity Canine, Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122920

 
life technologies
LS15095941
25 tests Each for $158.05

5014087

 
life technologies
LS15095942
100 tests Each for $375.55
Description & Specifications

Specifications

Antigen CD95 (APO-1/Fas)
Clone DX2
Host Species Mouse
Species Reactivity Canine, Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE-Cy5
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The DX2 monoclonal antibody reacts with human CD95 (Fas, Apo-1), a 40-50kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. DX2 does not block binding of EOS9.1, another antibody specific for human CD95.

The DX2 monoclonal is reported to recognize dog/canine CD95.