missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD95 (APO-1/Fas) Mouse anti-Human, PE, Clone: DX2, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $361.68

Specifications

Antigen CD95 (APO-1/Fas)
Clone DX2
Host Species Murine
Applications Flow Cytometry
Regulatory Status RUO
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50245282 Life Technologies
LS12095941
25 Tests Each for $157.08
50245283 Life Technologies
LS12095942
100 Tests Each for $361.68
Specifications

Description

The DX2 monoclonal antibody reacts with human CD95 (Fas, Apo-1), a 40-50kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. DX2 does not block binding of EOS9.1, another antibody specific for human CD95.

The DX2 monoclonal is reported to recognize dog/canine CD95.
Description & Specifications

Specifications

CD95 (APO-1/Fas)
Murine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
DX2
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test