missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD95 (APO-1/Fas) Mouse anti-Human, eFluor 450, Clone: DX2, eBioscience™

Mouse Monoclonal Antibody

$176.90 - $413.25

Specifications

Antigen CD95 (APO-1/Fas)
Clone DX2
Host Species Murine
Species Reactivity Canine, Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016316 Life Technologies
LS48095941
25 Tests Each for $176.90
5016317 Life Technologies
LS48095942
100 Tests Each for $413.25
Specifications

Description

The DX2 monoclonal antibody reacts with human CD95 (Fas, Apo-1), a 40-50kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. DX2 does not block binding of EOS9.1, another antibody specific for human CD95.

The DX2 monoclonal is reported to recognize dog/canine CD95.
Description & Specifications

Specifications

CD95 (APO-1/Fas)
Murine
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test
DX2
Canine, Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer