missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD95 (APO-1/Fas) Mouse anti-Human, Clone: EOS9.1, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$130.68 - $473.88

Specifications

Antigen CD95 (APO-1/Fas)
Clone EOS9.1
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry,Functional Assay
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014418 Life Technologies
LS16095881
50μg Each for $130.68
5014419 Life Technologies
LS16095885
500μg Each for $473.88
Specifications

Description

The EOS9.1 monoclonal antibody reacts with human CD95 (Fas, Apo-1), a 40-50kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, activated lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. EOS9.1 does not block binding of DX2, another antibody specific for human CD95.
Description & Specifications

Specifications

CD95 (APO-1/Fas)
Murine
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL
EOS9.1
Human
RUO
Purified
Primary
aqueous buffer, no sodium azide