Please login to your online account to display your discounted pricing

CD95 (APO-1/Fas) Mouse anti-Human, Clone: DX2, eBioscience™

Mouse Monoclonal Antibody

$129.05 - $245.05

Specifications

Antigen CD95 (APO-1/Fas)
Clone DX2
Host Species Murine
Species Reactivity Canine, Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012766

 
life technologies
LS14095980
25μg Each for $129.05

5012767

 
life technologies
LS14095982
100μg Each for $245.05
Description & Specifications

Specifications

Antigen CD95 (APO-1/Fas)
Clone DX2
Host Species Murine
Species Reactivity Canine, Human
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The DX2 monoclonal antibody reacts with human CD95 (Fas, Apo-1), a 40-50kDa member of the TNFR superfamily. CD95 is expressed by a broad range of hematopoietic and non-hematopoietic cells including monocytes, neutrophils, lymphocytes and fibroblasts. Interaction of CD95 on mature lymphocytes with its ligand (FasL) induces apoptosis and is thought to be important in peripheral tolerance. DX2 does not block binding of EOS9.1, another antibody specific for human CD95.