Please login to your online account to display your discounted pricing

CD94 Rat anti-Mouse, FITC, Clone: 18d3, eBioscience™

Rat Monoclonal Antibody

$100.05 - $230.55

Specifications

Antigen CD94
Clone 18d3
Host Species Rat
Gene Alias KLRD1, KP43
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509702

 
life technologies
LS11094181
50μg Each for $100.05

509703

 
life technologies
LS11094182
100μg Each for $230.55
Description & Specifications

Specifications

Antigen CD94
Clone 18d3
Host Species Rat
Gene Alias KLRD1, KP43
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 18d3 monoclonal antibody reacts with mouse CD94, a 70kDa type II transmembrane glycoprotein. CD94 belongs to the C-type lectin superfamily and is present as a heterodimer with NKG2 on the surface. CD94 is expressed by NK cells, a subset of T cells, and NKT cells and plays an important role in adhesion and activation of NK cell lineage. DBA/2J mice are naturally CD94-deficient and do not express cell-surface CD94/NKG2A receptors, even on neonatal NK cells.