missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD94 Mouse anti-Human, FITC, Clone: DX22, eBioscience™

Mouse Monoclonal Antibody

$117.48 - $236.28

Specifications

Antigen CD94
Clone DX22
Host Species Murine
Gene Alias KLRD1, KP43
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122083 Life Technologies
LS11094942
100 Tests Each for $236.28
509705 Life Technologies
LS11094941
25 Tests Each for $117.48
Specifications

Description

The DX22 monoclonal antibody reacts with human CD94, a 70kDa type II transmembrane glycoprotein. CD94 belongs to the C-type lectin superfamily and is present as a heterodimer with NKG2 on the surface. CD94 is expressed by NK cells, a subset of gd T cells, and NKT cells and plays an important role in adhesion and activation of NK cell lineage.
Description & Specifications

Specifications

CD94
Murine
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
DX22
KLRD1, KP43
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer