missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD93 (AA4.1) Rat anti-Mouse, APC, Clone: AA4.1, eBioscience™

Rat Monoclonal Antibody

$200.64 - $469.92

Specifications

Antigen CD93 (AA4.1)
Clone AA4.1
Host Species Rat
Gene Alias C1qRp, Early B lineage antigen
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5015236 Life Technologies
LS17589281
50μg Each for $200.64
5015237 Life Technologies
LS17589282
100μg Each for $335.28
5015238 Life Technologies
LS17589283
200μg Each for $469.92
Specifications

Description

The AA4.1 monoclonal antibody reacts with mouse C1qRp, an approximately 130-140kDa C-type lectin-like type I transmembrane protein. The AA4.1 antigen was originally identified as an antigen expressed on early stages of B cell development in bone marrow. Adult spleen and fetal liver also have detectable numbers of AA4.1+ cells. In combination with other markers of hematopoietic progenitor cells such as Thy-1, Sca-1, c-Kit, CD43, and CD24, the bone marrow Lymphoid-Committed Progenitors (CLP) can be segregated into more primitive and more differentiated subsets based on expression of AA4.1. Correlated expression of surface IgM (sIgM), CD23, and AA4.1 antigen has also been used to define three nonproliferative subpopulations of immature/transitional peripheral B cells designated: T1 (AA4.1+/CD23-/sIgMhi), T2 (AA4.1+/CD23+/sIgMhi), and T3 (AA4.1+/CD23+/sIgMlo). AA4.1 is also reported to be expressed in cytoplasmic vesicles in endothelial cells, megakaryoblasts, and platelets. It is reported that monoclonal antibodies AA4.1 and 493 recognize different epitopes of the same molecule.
Description & Specifications

Specifications

CD93 (AA4.1)
Rat
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL
AA4.1
C1qRp, Early B lineage antigen
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer