Please login to your online account to display your discounted pricing

CD90 (Thy-1) Rat anti-Mouse, Clone: G7, eBioscience™

Rat Monoclonal Antibody

$100.05 - $274.05

Specifications

Antigen CD90 (Thy-1)
Clone G7
Host Species Rat
Gene Alias Thy1
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012743

 
life technologies
LS14090181
50μg Each for $100.05

5012744

 
life technologies
LS14090182
100μg Each for $172.55

5012745

 
life technologies
LS14090185
500μg Each for $274.05
Description & Specifications

Specifications

Antigen CD90 (Thy-1)
Clone G7
Host Species Rat
Gene Alias Thy1
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The G7 monoclonal antibody reacts with mouse CD90 also known as Thy-1, a GPI-linked membrane molecule. CD90 is expressed by early hematopoietic cells in the bone marrow, thymocytes and mature T cells as well as neurons. CD90 is involved in regulation of adhesion and signal transduction by T cells. G7 is reported to induce TCR-mediated proliferation of T cell and apoptosis of thymocytes.