missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD90 (Thy-1) Rat anti-Canine, PE, Clone: YKIX337.217, eBioscience™

Rat Monoclonal Antibody

$130.68 - $328.68

Specifications

Antigen CD90 (Thy-1)
Clone YKIX337.217
Host Species Rat
Gene Alias Thy1
Species Reactivity Canine
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010980 Life Technologies
LS12590041
25 Tests Each for $130.68
5010981 Life Technologies
LS12590042
100 Tests Each for $328.68
Specifications

Description

This YKIX337.217 monoclonal antibody reacts with canine CD90, which is also known as Thy-1. This 25-37kDa N-glycoylsated cell adhesion molecule is expressed on thymocytes, peripheral T cells, some B cells, monocytes, and granulocytes albeit at a lower level. CD90 is also expressed on cells in the bone marrow, brain, and kidney, as well as on mesenchymal stem cells.

This monoclonal antibody has been reported to deplete cells in vitro and in vivo.
Description & Specifications

Specifications

CD90 (Thy-1)
Rat
Canine
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
YKIX337.217
Thy1
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test