missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD90 (Thy-1) Mouse anti-Human, Clone: eBio5E10 (5E10), eBioscience™

Mouse Monoclonal Antibody

$117.48 - $249.48

Specifications

Antigen CD90 (Thy-1)
Clone eBio5E10 (5E10)
Host Species Murine
Gene Alias Thy1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012752 Life Technologies
LS14090980
25μg Each for $117.48
5012753 Life Technologies
LS14090982
100μg Each for $249.48
Specifications

Description

The eBio5E10 monoclonal antibody reacts with human CD90, also known as Thy-1 (thymus cell antigen-1). CD90 is a 25-35 kD receptor expressed on thymocytes, CD34+ prothymocytes, hematopoietic stem cells, neurons, a small subset of human fetal liver cells, cord blood cells, and bone marrow cells. CD90 is expressed on a subset of immature, CD34+ cells and a distinct subset of mature CD34- cells that are CD3+CD4+. The CD90+CD34+ population is enriched for cells capable of long-term culture. CD90 is involved in regulation of adhesion and signal transduction by T cells.
Description & Specifications

Specifications

CD90 (Thy-1)
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBio5E10 (5E10)
Thy1
Flow Cytometry,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL