missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD90 (Thy-1) Mouse anti-Human, Biotin, Clone: eBio5E10 (5E10), eBioscience™

Mouse Monoclonal Antibody

$130.68 - $289.08

Specifications

Antigen CD90 (Thy-1)
Clone eBio5E10 (5E10)
Host Species Murine
Gene Alias Thy1
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011666 Life Technologies
LS13090980
25μg Each for $130.68
5011667 Life Technologies
LS13090982
100μg Each for $289.08
Specifications

Description

The eBio5E10 monoclonal antibody reacts with human CD90, also known as Thy-1 (thymus cell antigen-1). CD90 is a 25-35 kD receptor expressed on thymocytes, CD34+ prothymocytes, hematopoietic stem cells, neurons, a small subset of human fetal liver cells, cord blood cells, and bone marrow cells. CD90 is expressed on a subset of immature, CD34+ cells and a distinct subset of mature CD34- cells that are CD3+CD4+. The CD90+CD34+ population is enriched for cells capable of long-term culture. CD90 is involved in regulation of adhesion and signal transduction by T cells.
Description & Specifications

Specifications

CD90 (Thy-1)
Murine
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
eBio5E10 (5E10)
Thy1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer