Please login to your online account to display your discounted pricing

CD90.2 (Thy-1.2) Rat anti-Mouse, PerCP-eFluor 710, Clone: 30-H12, eBioscience™

Rat Monoclonal Antibody

$129.05 - $245.05

Specifications

Antigen CD90.2 (Thy-1.2)
Clone 30-H12
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015993

 
life technologies
LS46090380
25μg Each for $129.05

5015994

 
life technologies
LS46090382
100μg Each for $245.05
Description & Specifications

Specifications

Antigen CD90.2 (Thy-1.2)
Clone 30-H12
Host Species Rat
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 30-H12 monoclonal antibody reacts with mouse CD90.2, also known as Thy-1.2, a GPI-linked membrane molecule. CD90.2 is expressed by mouse thymocytes and mature T cells as well as neurons in CD90.2-expressing mouse strains. These strains include BALB/c, CBA, C3H, C57BL/6, C58/, SJL and others. Cells from CD90.1-expressing strains including PL and AKR do not stain with 30-H12. CD90 is involved in regulation of adhesion and signal transduction by T cells.