missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD90.2 (Thy-1.2) Rat anti-Mouse, PE-Cy7, Clone: 53-2.1, eBioscience™

Rat Monoclonal Antibody

$223.08 - $368.28

Specifications

Antigen CD90.2 (Thy-1.2)
Clone 53-2.1
Host Species Rat
Gene Alias Thy1.2
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123313 Life Technologies
LS25090282
100μg Each for $368.28
5015536 Life Technologies
LS25090281
50μg Each for $223.08
Specifications

Description

The 53-2.1 monoclonal antibody reacts with mouse CD90.2 also known as Thy-1.2, a GPI-linked membrane molecule. CD90.2 is expressed by mouse thymocytes and mature T cells as well as neurons in CD90.2-expressing mouse strains. These strains include BALB/c, CBA, C3H, C57BL/6, C58/, SJL and others. Cells from CD90.1-expressing strains including PL and AKR do not stain with 53-2.1. CD90 is involved in regulation of adhesion and signal transduction by T cells.
Description & Specifications

Specifications

CD90.2 (Thy-1.2)
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
53-2.1
Thy1.2
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL