missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD90.2 (Thy-1.2) Rat anti-Mouse, Clone: 30-H12, Functional Grade, eBioscience™

Rat Monoclonal Antibody, Functional Grade

$117.48 - $216.05

Specifications

Antigen CD90.2 (Thy-1.2)
Clone 30-H12
Host Species Rat
Gene Alias Thy1.2
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014415 Life Technologies
LS16090381
50μg Each for $117.48
5014416 Life Technologies
LS16090382
100μg Each for $216.05
Specifications

Description

The 30-H12 monoclonal antibody reacts with mouse CD90.2, also known as Thy-1.2, a GPI-linked membrane molecule. CD90.2 is expressed by mouse thymocytes and mature T cells as well as neurons in CD90.2-expressing mouse strains. These strains include BALB/c, CBA, C3H, C57BL/6, C58/, SJL and others. Cells from CD90.1-expressing strains including PL and AKR do not stain with 30-H12. CD90 is involved in regulation of adhesion and signal transduction by T cells.
Description & Specifications

Specifications

CD90.2 (Thy-1.2)
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, no sodium azide
30-H12
Thy1.2
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL