Please login to your online account to display your discounted pricing

CD90.2 (Thy-1.2) Rat anti-Mouse, Clone: 30-H12, eBioscience™

Rat Monoclonal Antibody

$114.55 - $230.55

Specifications

Antigen CD90.2 (Thy-1.2)
Clone 30-H12
Host Species Rat
Gene Alias Thy1.2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012749

 
life technologies
LS14090381
50μg Each for $114.55

5012750

 
life technologies
LS14090382
100μg Each for $172.55

5012751

 
life technologies
LS14090385
500μg Each for $230.55
Description & Specifications

Specifications

Antigen CD90.2 (Thy-1.2)
Clone 30-H12
Host Species Rat
Gene Alias Thy1.2
Species Reactivity Mouse
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 30-H12 monoclonal antibody reacts with mouse CD90.2, also known as Thy-1.2, a GPI-linked membrane molecule. CD90.2 is expressed by mouse thymocytes and mature T cells as well as neurons in CD90.2-expressing mouse strains. These strains include BALB/c, CBA, C3H, C57BL/6, C58/, SJL and others. Cells from CD90.1-expressing strains including PL and AKR do not stain with 30-H12. CD90 is involved in regulation of adhesion and signal transduction by T cells.