Please login to your online account to display your discounted pricing

CD90.2 (Thy-1.2) Rat anti-Mouse, Biotin, Clone: 53-2.1, eBioscience™

Rat Monoclonal Antibody

$85.55 - $274.05

Specifications

Antigen CD90.2 (Thy-1.2)
Clone 53-2.1
Host Species Rat
Gene Alias Thy1.2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011660

 
affymetrix
13090281
50μg Each for $85.55

5011661

 
affymetrix
13090282
100μg Each for $114.55

5011662

 
affymetrix
13090285
500μg Each for $274.05
Description & Specifications

Specifications

Antigen CD90.2 (Thy-1.2)
Clone 53-2.1
Host Species Rat
Gene Alias Thy1.2
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 53-2.1 monoclonal antibody reacts with mouse CD90.2 also known as Thy-1.2, a GPI-linked membrane molecule. CD90.2 is expressed by mouse thymocytes and mature T cells as well as neurons in CD90.2-expressing mouse strains. These strains include BALB/c, CBA, C3H, C57BL/6, C58/, SJL and others. Cells from CD90.1-expressing strains including PL and AKR do not stain with 53-2.1. CD90 is involved in regulation of adhesion and signal transduction by T cells.