Please login to your online account to display your discounted pricing

CD90.1 (Thy-1.1) Mouse anti-Mouse, Rat, FITC, Clone: HIS51, eBioscience™

Mouse Monoclonal Antibody

$85.55 - $419.05

Specifications

Antigen CD90.1 (Thy-1.1)
Clone HIS51
Host Species Mouse
Gene Alias Thy1.1
Species Reactivity Mouse, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

509692

 
affymetrix
11090085
500μg Each for $419.05

501129595

 
affymetrix
11090081
50μg Each for $85.55
Description & Specifications

Specifications

Antigen CD90.1 (Thy-1.1)
Clone HIS51
Host Species Mouse
Gene Alias Thy1.1
Species Reactivity Mouse, Rat
Applications Flow Cytometry
Regulatory Status RUO
Conjugate FITC
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The HIS51 monoclonal antibody reacts with rat CD90 and cross-reacts with mouse CD90.1, a GPI-linked membrane molecule. In the rat, CD90 is expressed by hematopoietic stem cells, immature B cells, thymocytes, recent thymic emigrants, neurons, inflammed endothelia and other cell types. In the CD90.1-expressing mouse strains, PL and AKR, CD90 is expressed by early hematopoietic cells in the bone marrow, thymocytes and mature T cells.