missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD9 Rat anti-Mouse, Clone: eBioKMC8 (KMC8), eBioscience™

Rat Monoclonal Antibody

$85.55 - $361.05

Specifications

Antigen CD9
Clone eBioKMC8 (KMC8)
Host Species Rat
Gene Alias MIC3, Tspan29
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012400 Life Technologies
LS14009181
50μg Each for $85.55
5012401 Life Technologies
LS14009182
100μg Each for $158.05
5012402 Life Technologies
LS14009185
500μg Each for $361.05
Specifications

Description

The eBioKMC8 monoclonal antibody reacts with mouse CD9, a 24kDa member of the transmembrane 4 superfamily. This family is characterized by the presence of four hydrophobic domains spanning the cell membrane and short N- and C-terminal cytoplasmic domains. CD9 is expressed by several cell types including monocytes, macrophages, platelets, early B cells, activated B and T cells, dendritic cells, eosinophils, basophils, endothelial cells, myoblasts and neuroblasts. On T cells, CD9 functions as a co-stimulatory molecule on naïve T cells. Furthermore, CD9 is expressed in oocytes, and CD9-deficiency results in sterility caused by defective gamete fusion. In mouse macrophages, CD9 functionally associates with FcγRs to modify signals for phagocytosis and inflammatory responses. In mouse B cells, it was discovered that CD9 is a marker for marginal zone B cells, B1 cells, and plasma cells. In dendritic cells, recently it was demonstrated that CD9 facilitates the association of heterologous MHC II molecules. The level of CD9 expression is subject to donor variability.
Description & Specifications

Specifications

CD9
Rat
Mouse
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioKMC8 (KMC8)
MIC3, Tspan29
Flow Cytometry,Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL