Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD9/Motility-Related Protein-1 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD9/Motility-Related Protein-1 Ab-1, Mouse Monoclonal Antibody.

$295.47 - $1,283.29

Specifications

Antigen CD9/Motility-Related Protein-1 Ab-1
Clone 72F6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS1077R7

 
thermo scientific™ lab vision™
MS1077R7
7mL Each for $865.18

MS1077S

 
thermo scientific™ lab vision™
MS1077S
1mL Each for $1,283.29

MS1077S0

 
thermo scientific™ lab vision™
MS1077S0
100µL Each for $295.47

MS1077S1

 
thermo scientific™ lab vision™
MS1077S1
500µL Each for $874.69
Description & Specifications

Specifications

Antigen CD9/Motility-Related Protein-1 Ab-1
Clone 72F6
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Hematopoietic
Type Primary
Immunogen Recombinant protein encoding the major extracellular loop of the human CD9
Isotype IgG1

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD9/Motility-Related Protein-1 Ab-1, Mouse Monoclonal Antibody.

CD9 antigen is a glycoprotein expressed on the surface of developing B lymphocytes, platelets, monocytes, eosinophils, basophil, stimulated T lymphocytes and by neurons and glial cells in the peripheral nervous system. It belongs to a family of membrane proteins termed tetraspanins which transverse the membrane four times. In pre-B cells and platelets, CD9 antigen regulates cell activation and aggregation possibly through an association with the integrin CD41/CD61 (GPIIb/GPIIIa).

Host Species: Mouse

Clone: 72F6

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Epitope: Extracellular domain

Immunogen: Recombinant protein encoding the major extracellular loop of the human CD9

Molecular Weight: 24-27kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO