missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD8b Rat anti-Mouse, PE-Cy7, Clone: eBioH35-17.2 (H35-17.2), eBioscience™

Rat Monoclonal Antibody

$147.84 - $361.68

Specifications

Antigen CD8b
Clone eBioH35-17.2 (H35-17.2)
Host Species Rat
Gene Alias CD8 beta, Ly-3, Ly-C, Lyt-3
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123262 Life Technologies
LS25008380
25μg Each for $147.84
501123263 Life Technologies
LS25008382
100μg Each for $361.68
Specifications

Description

The eBioH35-17.2 monoclonal antibody reacts with the mouse CD8 beta molecule. The CD8 beta chain associates with the CD8 alpha chain to form the CD8 alpha/beta heterodimer expressed on the surface of a majority of thymocytes, and on peripheral cytotoxic alpha beta TCR T cells. CD8 binds to MHC class I and plays a role in T cell development and activation of mature T cells.
Description & Specifications

Specifications

CD8b
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
eBioH35-17.2 (H35-17.2)
CD8 beta, Ly-3, Ly-C, Lyt-3
Flow Cytometry
PE-Cy7
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL