Please login to your online account to display your discounted pricing

CD8b Rat anti-Mouse, eFluor 450, Clone: eBioH35-17.2 (H35-17.2), eBioscience™

Rat Monoclonal Antibody

$158.05 - $390.05

Specifications

Antigen CD8b
Clone eBioH35-17.2 (H35-17.2)
Host Species Rat
Gene Alias CD8 beta, Ly-3, Ly-C, Lyt-3
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124084

 
affymetrix
48008380
25μg Each for $158.05

501128793

 
affymetrix
48008382
100μg Each for $390.05
Description & Specifications

Specifications

Antigen CD8b
Clone eBioH35-17.2 (H35-17.2)
Host Species Rat
Gene Alias CD8 beta, Ly-3, Ly-C, Lyt-3
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 450
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The eBioH35-17.2 monoclonal antibody reacts with the mouse CD8 beta molecule. The CD8 beta chain associates with the CD8 alpha chain to form the CD8 alpha/beta heterodimer expressed on the surface of a majority of thymocytes, and on peripheral cytotoxic alpha beta TCR T cells. CD8 binds to MHC class I and plays a role in T cell development and activation of mature T cells.