Please login to your online account to display your discounted pricing

CD8a Rat anti-Mouse, PE, Clone: 53-6.7, eBioscience™

Rat Monoclonal Antibody

$85.55 - $535.05

Specifications

Antigen CD8a
Clone 53-6.7
Host Species Rat
Gene Alias CD8 alpha, Ly-2, Ly-35, Ly-B, Lyt-2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010183

 
life technologies
LS12008181
50μg Each for $85.55

5010186

 
life technologies
LS12008185
500μg Each for $535.05

501129416

 
life technologies
LS12008182
100μg Each for $129.05

501129660

 
life technologies
LS12008183
200μg Each for $216.05
Description & Specifications

Specifications

Antigen CD8a
Clone 53-6.7
Host Species Rat
Gene Alias CD8 alpha, Ly-2, Ly-35, Ly-B, Lyt-2
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 53-6.7 monoclonal antibody reacts with the mouse CD8a molecule. CD8a is an approximately 32-34kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha beta) or as a homodimer (CD8 alpha alpha). A majority of thymocytes and a subpopulation of mature alpha beta TCR T cells express CD8 alpha beta while gamma delta TCR T cells, a subpopulation of intestinal intraepithelial lymphocytes (IELs) and dendritic cells express CD8 alpha alpha. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells.