Please login to your online account to display your discounted pricing

CD8a Rat anti-Mouse, eVolve 655, Clone: 53-6.7, eBioscience™

Rat Monoclonal Antibody

$187.05 - $382.80

Specifications

Antigen CD8a
Clone 53-6.7
Host Species Rat
Gene Alias CD8 alpha, Ly-2, Ly-35, Ly-B, Lyt-2
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501125040

 
life technologies
LS86008141
25 Tests Each for $187.05

501125041

 
life technologies
LS86008142
100 Tests Each for $382.80
Description & Specifications

Specifications

Antigen CD8a
Clone 53-6.7
Host Species Rat
Gene Alias CD8 alpha, Ly-2, Ly-35, Ly-B, Lyt-2
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eVolve 655
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL

The 53-6.7 monoclonal antibody reacts with the mouse CD8a molecule. CD8a is an approximately 32-34kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha beta) or as a homodimer (CD8 alpha alpha). A majority of thymocytes and a subpopulation of mature alpha beta TCR T cells express CD8 alpha beta while gamma delta TCR T cells, a subpopulation of intestinal intraepithelial lymphocytes (IELs) and dendritic cells express CD8 alpha alpha. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells.