missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD8a Rat anti-Mouse, Biotin, Clone: 53-6.7, eBioscience™

Rat Monoclonal Antibody

$64.68 - $513.48

Specifications

Antigen CD8a
Clone 53-6.7
Host Species Rat
Gene Alias CD8 alpha, Ly-2, Ly-35, Ly-B, Lyt-2
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122502 Life Technologies
LS13008181
50μg Each for $64.68
5011408 Life Technologies
LS13008182
100μg Each for $91.08
5011409 Life Technologies
LS13008185
500μg Each for $289.08
5011410 Life Technologies
LS13008186
1mg Each for $513.48
Specifications

Description

The 53-6.7 monoclonal antibody reacts with the mouse CD8a molecule. CD8a is an approximately 32-34kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha beta) or as a homodimer (CD8 alpha alpha). A majority of thymocytes and a subpopulation of mature alpha beta TCR T cells express CD8 alpha beta while gamma delta TCR T cells, a subpopulation of intestinal intraepithelial lymphocytes (IELs) and dendritic cells express CD8 alpha alpha. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells.
Description & Specifications

Specifications

CD8a
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
53-6.7
CD8 alpha, Ly-2, Ly-35, Ly-B, Lyt-2
Flow Cytometry,Immunohistochemistry (Frozen)
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL