missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD8a Rat anti-Mouse, APC-eFluor 780, Clone: 53-6.7, eBioscience™

Rat Monoclonal Antibody

$129.05 - $288.55

Specifications

Antigen CD8a
Clone 53-6.7
Host Species Rat
Gene Alias CD8 alpha, Ly-2, Ly-35, Ly-B, Lyt-2
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016135 Life Technologies
LS47008180
25μg Each for $129.05
501129358 Life Technologies
LS47008182
100μg Each for $288.55
Specifications

Description

The 53-6.7 monoclonal antibody reacts with the mouse CD8a molecule. CD8a is an approximately 32-34kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha beta) or as a homodimer (CD8 alpha alpha). A majority of thymocytes and a subpopulation of mature alpha beta TCR T cells express CD8 alpha beta while gamma delta TCR T cells, a subpopulation of intestinal intraepithelial lymphocytes (IELs) and dendritic cells express CD8 alpha alpha. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells.
Description & Specifications

Specifications

CD8a
Rat
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
53-6.7
CD8 alpha, Ly-2, Ly-35, Ly-B, Lyt-2
Flow Cytometry
APC-eFluor 780
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
0.2mg/mL