Please login to your online account to display your discounted pricing

CD8a Mouse, eFluor 506, Clone: RPA-T8, eBioscience™

Mouse Monoclonal Antibody

$176.90 - $382.80

Specifications

Antigen CD8a
Clone RPA-T8
Host Species Mouse
Gene Alias CD8 alpha, leu-2a
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124927

 
life technologies
LS69008841
25 tests Each for $176.90

501124928

 
life technologies
LS69008842
100 tests Each for $382.80
Description & Specifications

Specifications

Antigen CD8a
Clone RPA-T8
Host Species Mouse
Gene Alias CD8 alpha, leu-2a
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 506
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.125μg)/test

The RPA-T8 monoclonal antibody reacts with the human CD8a molecule, an approximately 32-34kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T-cell development and activation of mature T cells.