missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD8a Mouse anti-Rat, FITC, Clone: OX8, eBioscience™

Mouse Monoclonal Antibody

$77.88 - $196.68

Specifications

Antigen CD8a
Clone OX8
Host Species Murine
Gene Alias CD8 alpha
Species Reactivity Rat
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122035 Life Technologies
LS11008482
100μg Each for $196.68
501122034 Life Technologies
LS11008480
25μg Each for $77.88
Specifications

Description

The OX8 monoclonal antibody reacts with rat CD8 alpha. CD8 alpha is a member of the immunoglobulin superfamily which heterodimerizes with CD8 beta to form the CD8 antigen expressed on CD4+CD8+, double-positive thymocytes, the majority of MHC class I-restricted peripheral T cells, and on some macrophages. CD8 binds to MHC class I expressed on the surface of antigen-presenting cells during antigen presentation, and participates in T-cell receptor signal transduction through association with the kinase Lck.
Description & Specifications

Specifications

CD8a
Murine
Rat
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
OX8
CD8 alpha
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL