missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD8a Mouse anti-Human, eFluor 450, Clone: SK1, eBioscience™

Mouse Monoclonal Antibody

$176.90 - $397.30

Specifications

Antigen CD8a
Clone SK1
Host Species Murine
Gene Alias CD8 alpha, leu-2a
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124085 Life Technologies
LS48008741
25 Tests Each for $176.90
501124086 Life Technologies
LS48008742
100 Tests Each for $397.30
Specifications

Description

The SK1 monoclonal antibody reacts with the human CD8a molecule, an approximately 32-34kDa cell surface receptor expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T-cell development and activation of mature T cells.
Description & Specifications

Specifications

CD8a
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
SK1
CD8 alpha, leu-2a
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test