missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD8a Mouse anti-Human, Clone: AMC908, eBioscience™

Mouse Monoclonal Antibody

$87.00 - $220.40

Specifications

Antigen CD8a
Clone AMC908
Host Species Murine
Gene Alias CD8 alpha, leu-2a
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122626 Life Technologies
LS14000880
25μg Each for $87.00
501122627 Life Technologies
LS14000882
100μg Each for $220.40
Specifications

Description

This AMC908 monoclonal antibody reacts with the human CD8a molecule, an approximately 32-34kDa cell surface receptor expressed either as a heterodimer with the CD8 β chain (CD8 αβ) or as a homodimer (CD8 αα). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T-cell development and activation of mature T cells.
Description & Specifications

Specifications

CD8a
Murine
Human
RUO
Purified
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
AMC908
CD8 alpha, leu-2a
Immunohistochemistry,Immunohistochemistry (Formalin/Paraffin),Western Blot
Unlabeled
Store at 2-8°C.
Monoclonal
0.5mg/mL