Please login to your online account to display your discounted pricing

CD8a Mouse anti-Human, Biotin, Clone: AMC908, eBioscience™

Mouse Monoclonal Antibody

$101.50 - $101.50

Specifications

Antigen CD8a
Clone AMC908
Host Species Murine
Gene Alias CD8 alpha, leu-2a
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122496

 
life technologies
LS13000880
25μg Each for $101.50
Description & Specifications

Specifications

Antigen CD8a
Clone AMC908
Host Species Murine
Gene Alias CD8 alpha, leu-2a
Species Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Western Blot
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This AMC908 monoclonal antibody reacts with the human CD8a molecule, an approximately 32-34kDa cell surface receptor expressed either as a heterodimer with the CD8 β chain (CD8 αβ) or as a homodimer (CD8 αα). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T-cell development and activation of mature T cells.