missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD86 (B7-2) Mouse anti-Human, Clone: IT2.2, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$143.88 - $460.68

Specifications

Antigen CD86 (B7-2)
Clone IT2.2
Host Species Murine
Gene Alias B72, B70
Applications Flow Cytometry,Functional Assay
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5014410 Life Technologies
LS16086981
50μg Each for $143.88
5014411 Life Technologies
LS16086985
500μg Each for $460.68
Specifications

Description

The IT2.2 monoclonal antibody reacts with human CD86, an ∽80kDa surface receptor also known as B7-2. CD86 and CD80 are members of the B7 family of costimulatory molecules. CD86 is expressed at low levels on B cells, macrophages, and dendritic cells and is upregulated on B cells through a variety of surface stimuli including the BCR complex, CD40 and some cytokine receptors. In addition to CD80 (B7-1), CD86 is a counter-receptor for the T cell surface molecules CD28 and CD152 (CTLA-4). The interaction of CD86 with its ligands plays a critical role in T-B crosstalk, T cell costimulation, autoantibody production and Th2-mediated Ig production. The kinetics of upregulation of CD86 upon stimulation supports its major contribution during the primary phase of an immune response.
Description & Specifications

Specifications

CD86 (B7-2)
Murine
Flow Cytometry,Functional Assay
Unlabeled
Store at 2-8°C.
Monoclonal
1mg/mL
IT2.2
B72, B70
RUO
Purified
Primary
aqueous buffer, no sodium azide