missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD85g (ILT7) Mouse anti-Human, APC, Clone: 17G10.2, eBioscience™

Mouse Monoclonal Antibody

$130.68 - $328.68

Specifications

Antigen CD85g (ILT7)
Clone eBio17G10.2 (17G10.2)
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501123129 Life Technologies
LS17517942
100 Tests Each for $328.68
5015191 Life Technologies
LS17517941
25 Tests Each for $130.68
Specifications

Description

The eBio17G10.2 monoclonal antibody reacts with human immunoglobulin-like transcript 7 (ILT7), also known as CD85g. ILT7 is part of the leukocyte immunoglobulin-like receptor (LIR) gene family and associates with FcεR1γ to form a functional complex which transduces ITAM-mediated signals that negatively modulates TLR-induced type I IFN production by human plasmacytoid dendritic cells. ILT7 is expressed exclusively on plasmacytoid dendritic cells and is absent in monocyte derived DCs. Treatment with IL-3 or CpG causes a decrease in expression.

Double staining of human PBMCs with eBio17G10.2 and anti-pDC marker BDCA2 yields virtually complete co-staining. Cross linking of ILT7 with eBio17G10.2 antibody causes reduced transcription of IFN-α1, IFN-α2, IFN-α4 and IFN-β.
Description & Specifications

Specifications

CD85g (ILT7)
Murine
Flow Cytometry
Allophycocyanin
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
eBio17G10.2 (17G10.2)
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer