missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD85d (ILT4) Rat anti-Human, FITC, Clone: 27D6, eBioscience™

Rat Monoclonal Antibody

$130.68 - $249.48

Specifications

Antigen CD85d (ILT4)
Clone 27D6
Host Species Rat
Gene Alias ILT-4, LIR2, LILRB2, MIR10
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509825 Life Technologies
LS11514841
25 Tests Each for $130.68
509826 Life Technologies
LS11514842
100 Tests Each for $249.48
Specifications

Description

The monoclonal antibody 27D6 recognizes the CD85d protein also known as ILT4, LIR-2. It is a member of the ILT (immunoglobulin-like transcript)/ LIR (leuokocyte Ig-like receptor) MIR (monocytes Ig-like receptor) family. CD85d is a 110kDa single transmembrane receptor containing 3 Ig-like domains and a long cytoplasmic domain containing ITIMs (immunoreceptor tyrosine-based inhibitory motifs). Expression is confined to monocytes, macrophages and dendritic cells. The ligand for CD85d is non-classical class I HLA-G, -B and G1. Upon ligand binding, tyrosine phosphatase SHP-1 interacts with the ITIM. Interaction of HLA-G with ILT4/CD85d is thought to contribute to tolerance during fetal implantation. Additionally it is the levels of ILT3 and ILT4 on DCs that suggests a role in organ allograft rejection.

The monoclonal antibody 27D6 has been shown to block binding of ligand (HLA-G tetramer).
Description & Specifications

Specifications

CD85d (ILT4)
Rat
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
27D6
ILT-4, LIR2, LILRB2, MIR10
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.5μg)/test