missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD83 Rat anti-Mouse, FITC, Clone: Michel-17 (Michel17), eBioscience™

Rat Monoclonal Antibody

$130.68 - $262.68

Specifications

Antigen CD83
Clone Michel-17 (Michel17)
Host Species Rat
Gene Alias HB15
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509680 Life Technologies
LS11083180
25μg Each for $130.68
509681 Life Technologies
LS11083182
100μg Each for $262.68
Specifications

Description

The Michel-17 monoclonal antibody reacts with mouse CD83, a 45kDa cell surface glycoprotein and a member of the Ig superfamily. The mouse CD83 antigen is expressed predominantly on mature DC and activated lymphocytes. Cross-linking of CD83 with Michel-17 on DC or activated T cells does not induce any activation signal. CD83 plays an important role in T cell development through interaction with its ligand. CD83-Ig protein has revealed the presence of a CD83 ligand expressed mainly by B220+ cells in mouse spleen.
Description & Specifications

Specifications

CD83
Rat
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.5mg/mL
Michel-17 (Michel17)
HB15
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer