Please login to your online account to display your discounted pricing

CD83 Rat anti-Mouse, Biotin, Clone: Michel-17 (Michel17), eBioscience™

Rat Monoclonal Antibody

$129.05 - $303.05

Specifications

Antigen CD83
Clone Michel-17 (Michel17)
Host Species Rat
Gene Alias HB15
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011643

 
life technologies
LS13083180
25μg Each for $129.05

5011644

 
life technologies
LS13083182
100μg Each for $303.05
Description & Specifications

Specifications

Antigen CD83
Clone Michel-17 (Michel17)
Host Species Rat
Gene Alias HB15
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The Michel-17 monoclonal antibody reacts with mouse CD83, a 45kDa cell surface glycoprotein and a member of the Ig superfamily. The mouse CD83 antigen is expressed predominantly on mature DC and activated lymphocytes. Cross-linking of CD83 with Michel-17 on DC or activated T cells does not induce any activation signal. CD83 plays an important role in T cell development through interaction with its ligand. CD83-Ig protein has revealed the presence of a CD83 ligand expressed mainly by B220+ cells in mouse spleen.