missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD83 Mouse anti-Human, PE, Clone: HB15e, eBioscience™

Mouse Monoclonal Antibody

$170.28 - $369.60

Specifications

Antigen CD83
Clone HB15e
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5010491 Life Technologies
LS12083941
25 Tests Each for $170.28
5010492 Life Technologies
LS12083942
100 Tests Each for $369.60
Specifications

Description

The HB15e monoclonal antibody reacts with human CD83, a 45kDa transmembrane glycoprotein. CD83, a member of the Ig superfamily, is expressed on cultured dendritic cells, interdigitating, follicular, and circulating dendritic cells as well as some proliferating lymphocytes, and human cell lines express this antigen. While the function of CD83 is unclear, it can serve as a useful marker for mature human blood dendritic cells.
Description & Specifications

Specifications

CD83
Murine
Flow Cytometry
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.25μg)/test
HB15e
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer