Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD81/TAPA-1 Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, flow cytometry and functional study experiments.

Manufacturer: thermo scientific™ lab vision™  MS502P1ABX

 View more versions of this product

Catalog No. MS502P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Immunogen B cell line derived from a Burkitt lymphoma
Host Species Mouse
Antigen CD81/TAPA-1 Ab-1
Format Concentrated
Quantity 100µL
Clone 1.3.3.22
Cross Reactivity Human
Isotype IgG2a kappa
Regulatory Status RUO
Conjugate Unlabeled
Concentration 1mg/mL
Type Primary
Classification Monoclonal
Research Discipline Signal Transduction

Ensure accurate, reproducible results in immunohistochemistry, immunofluorescence, flow cytometry and functional study experiments with Thermo Scientific™ CD81/TAPA-1 Ab-1, Mouse Monoclonal Antibody.

CD81 (target of anti-proliferative antibody-1 (TAPA-1) has a very broad cellular distribution, being expressed on T and B lymphocytes, NK cells, thymocytes, eosinophils, fibroblasts, epithelial and endothelial cells. Neutrophils, erythrocytes and platelets are negative, while monocytes are variably positive. CD81 is a member of a family of tetraspans transmembrane proteins, including CD9, CD37, CD53, CD63, and CD82. It associates with CD19, CD21, Leu 13, and integrins on cell membrane and is involved in signal transduction in B lymphocyte development and cell adhesion. CD81 also acts as a receptor for the envelope protein E2 of chronic hepatitis C virus. Antibodies to CD81 have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas.

Host Species: Mouse

Clone: 1.3.3.22

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: B-cell line derived from a Burkitt lymphoma

Molecular Weight: 24 kDa

Positive Control: Daudi, Raji, U266, HUT-78, HUT-102, Jurkat, HL-60, KG1a, A293, A431, MG63, and human lymphocytes; Tonsil/lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Acetone-fixed frozen)
  • Immunofluorescence
  • Flow Cytometry
  • Functional Study

Note:

USA: RUO; Int'l: RUO