missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD81 Armenian Hamster anti-Mouse, Biotin, Clone: Eat-2 (EAT2), eBioscience™

Armenian Hamster Monoclonal Antibody

$117.48 - $259.55

Specifications

Antigen CD81
Clone Eat-2 (EAT2)
Host Species Armenian Hamster
Gene Alias TAPA-1, TAPA1
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5011640 Life Technologies
LS13081181
50μg Each for $117.48
5011641 Life Technologies
LS13081182
100μg Each for $259.55
Specifications

Description

The Eat-2 (EAT2) monoclonal antibody reacts with mouse CD81, a molecule also known as TAPA-1. CD81 is expressed on CD4+CD8+ thymocytes and broadly in the periphery with high level expressed by B cells, NK cells, macrophages and dendritic cells and lower level on resting T cells. Activation of T cells induces upregulation of CD81. CD81 is involved in homotypic adhesion of B and T cells as well as costimulation of T cells. The Eat-2 antibody is reported to cross-react with rat CD81.
Description & Specifications

Specifications

CD81
Armenian Hamster
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
Eat-2 (EAT2)
TAPA-1, TAPA1
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer