Please login to your online account to display your discounted pricing

CD80 (B7-1) Mouse anti-Human, PE, Clone: 2D10.4, eBioscience™

Mouse Monoclonal Antibody

$130.50 - $321.90

Specifications

Antigen CD80 (B7-1)
Clone 2D10.4
Host Species Mouse
Gene Alias B71, BB1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5010487

 
life technologies
LS12080941
25 tests Each for $130.50

5010488

 
life technologies
LS12080942
100 tests Each for $321.90
Description & Specifications

Specifications

Antigen CD80 (B7-1)
Clone 2D10.4
Host Species Mouse
Gene Alias B71, BB1
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

The 2D10.4 monoclonal antibody reacts with human CD80 (B7-1), a 60kDa member of the Ig superfamily. CD80 is expressed by activated B cells, macrophages and dendritic cells. In addition, activated T cells express this antigen. CD80 has high affinity for binding to two T cell surface antigens, CD28 and CD152 (CTLA-4). The interaction of CD28 and CD152 with CD80 is crucial in T-B cell communication leading to activation of T and B cells, respectively.