Please login to your online account to display your discounted pricing

CD80 (B7-1) Mouse anti-Human, Clone: 2D10.4, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$158.05 - $462.55

Specifications

Antigen CD80 (B7-1)
Clone 2D10.4
Host Species Mouse
Gene Alias B71, BB1
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014392

 
affymetrix
16080981
50μg Each for $158.05

5014393

 
affymetrix
16080985
500μg Each for $462.55
Description & Specifications

Specifications

Antigen CD80 (B7-1)
Clone 2D10.4
Host Species Mouse
Gene Alias B71, BB1
Species Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The 2D10.4 monoclonal antibody reacts with human CD80 (B7-1), a 60kDa member of the Ig superfamily. CD80 is expressed by activated B cells, macrophages and dendritic cells. In addition, activated T cells express this antigen. CD80 has high affinity for binding to two T cell surface antigens, CD28 and CD152 (CTLA-4). The interaction of CD28 and CD152 with CD80 is crucial in T-B cell communication leading to activation of T and B cells, respectively.