Please login to your online account to display your discounted pricing

CD80 (B7-1) Armenian Hamster anti-Mouse, Clone: 16-10A1, Functional Grade, eBioscience™

Armenian Hamster Monoclonal Antibody, Functional Grade

$230.55 - $404.55

Specifications

Antigen CD80 (B7-1)
Clone 16-10A1
Host Species Armenian Hamster
Gene Alias B71, Ly-53
Species Reactivity Canine, Mouse, Pig
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014389

 
affymetrix
16080182
100μg Each for $230.55

5014390

 
affymetrix
16080185
500μg Each for $404.55
Description & Specifications

Specifications

Antigen CD80 (B7-1)
Clone 16-10A1
Host Species Armenian Hamster
Gene Alias B71, Ly-53
Species Reactivity Canine, Mouse, Pig
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The 16-10A1 monoclonal antibody reacts with mouse CD80 (B7-1), a 55kDa member of the Ig superfamily. CD80 is expressed by macrophages, dendritic cells and activated B cells. In addition, activated T cells express this antigen. CD80 has high affinity for binding to two T cell surface antigens, CD28 and CD152 (CTLA-4). The interaction of CD28 and CD152 with CD80 is crucial in T-B cell communication leading to activation of T and B cells, respectively.