Please login to your online account to display your discounted pricing

CD8 Rat anti-Canine, APC, Clone: YCATE55.9, eBioscience™

Rat Monoclonal Antibody

$158.05 - $382.80

Specifications

Antigen CD8
Clone YCATE55.9
Host Species Rat
Gene Alias CD8 alpha
Species Reactivity Canine
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5015185

 
life technologies
LS17508041
25 Tests Each for $158.05

5015186

 
life technologies
LS17508042
100 Tests Each for $382.80
Description & Specifications

Specifications

Antigen CD8
Clone YCATE55.9
Host Species Rat
Gene Alias CD8 alpha
Species Reactivity Canine
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Allophycocyanin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.5μg)/test

This YCATE55.9 monoclonal antibody reacts with canine CD8 alpha. This cell surface receptor is expressed either as a heterodimer with the CD8 beta chain (CD8 alpha/beta) or as a homodimer (CD8 alpha/alpha). In dogs, CD8 alpha is expressed on thymocytes, a subset of mature T cells, NK cells, and intestinal intraepithelial lymphocytes (IELs). CD8 binds to MHC class I and plays a role in thymocyte development and mature T cell activation.

This monoclonal antibody has been reported to inhibit cytotoxic T cell generation and function in vitro. In addition, this antibody has been reported to be immunosuppressive in vivo.