Please login to your online account to display your discounted pricing

CD8 Mouse anti-Human, Biotin, Clone: OKT8 (OKT-8), eBioscience™

Mouse Monoclonal Antibody

$114.55 - $259.55

Specifications

Antigen CD8
Clone OKT8 (OKT-8)
Host Species Murine
Gene Alias CD8 alpha, leu-2a
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011414

 
life technologies
LS13008680
25μg Each for $114.55

5011415

 
life technologies
LS13008682
100μg Each for $259.55
Description & Specifications

Specifications

Antigen CD8
Clone OKT8 (OKT-8)
Host Species Murine
Gene Alias CD8 alpha, leu-2a
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The OKT8 monoclonal antibody reacts with the human CD8a molecule, an approximately 32-34kDa cell surface receptor expressed either as a heterodimer with the CD8 β chain (CD8 αβ) or as a homodimer (CD8 αα). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class I and through its association with protein tyrosine kinase p56lck plays a role in T-cell development and activation of mature T cells. Preliminary testing indicates that OKT8 and two other mouse anti-human CD8 antibodies (clone RPA-T8, Cat. No.14-0088 and clone HIT8a, Cat. No.14-0089) do not compete with each other for binding to human peripheral blood leukocytes by flow cytometric analysis, suggesting that they do not bind to similar epitopes or block each other by steric hindrance.