missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

CD79b Mouse anti-Human, FITC, Clone: CB3-1, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $332.05

Specifications

Antigen CD79b
Clone CB3-1
Host Species Murine
Gene Alias Ig-beta, B cell Receptor
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
509670 Life Technologies
LS11079341
25 Tests Each for $172.55
509671 Life Technologies
LS11079342
100 Tests Each for $332.05
Specifications

Description

This CB3-1 monoclonal antibody reacts with human CD79b, the B cell antigen receptor Ig-beta chain. Together with the Ig-alpha chain, also known as CD79a, this cell surface molecule forms a heterodimeric complex that associates with membrane immunoglobulin (mIgM). CD79b expression is exclusive to B lymphocytes and B cell lymphomas. Depending on the maturation state of the B cell, CD79b can be expressed either in the cytoplasm or on the cell surface. The CD79 receptor complex triggers numerous signaling pathways to mediate B cell development, maintenance, and activation.
Description & Specifications

Specifications

CD79b
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CB3-1
Ig-beta, B cell Receptor
Flow Cytometry
FITC
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (1μg)/test