missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ CD79a/mb-1 (B-Cell Marker), Rabbit Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD79a/mb-1 (B-Cell Marker), Rabbit Monoclonal Antibody.

$303.71 - $1,245.59

Specifications

Antigen CD79a/mb-1 (B-Cell Marker)
Clone SP18
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
RM9118R7 Thermo Scientific™ Lab Vision™
RM9118R7
7mL Each for $644.59
RM9118S Thermo Scientific™ Lab Vision™
RM9118S
1mL Each for $1,245.59
RM9118S0 Thermo Scientific™ Lab Vision™
RM9118S0
100μL Each for $303.71
RM9118S1 Thermo Scientific™ Lab Vision™
RM9118S1
500μL Each for $891.92
Specifications

Description

Obtain accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ CD79a/mb-1 (B-Cell Marker), Rabbit Monoclonal Antibody, part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.

A disulphide-linked heterodimer, consisting of CD79a/mb-1 and CD79b/B29 polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas.

Host Species: Rabbit

Clone: SP18

Species Reactivity: Human. Does not react with mouse, rat, canine, monkey and opossum

Epitope: N-terminus

Immunogen: Synthetic peptide derived from N-terminal region of human CD79a protein

Molecular Weight: 50 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Description & Specifications

Specifications

CD79a/mb-1 (B-Cell Marker)
Immunohistochemistry (Paraffin)
Unlabeled
IVD
Primary
SP18
Monoclonal
Rabbit
Cancer and Tumor Biology
Synthetic peptide derived from N-terminal region of human CD79a protein