Please login to your online account to display your discounted pricing

CD75 Mouse anti-Human, eFluor 660, Clone: LN-1, eBioscience™

Mouse Monoclonal Antibody

$143.55 - $361.05

Specifications

Antigen CD75
Clone LN-1
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124285

 
life technologies
LS50075941
25 tests Each for $143.55

501124286

 
life technologies
LS50075942
100 tests Each for $361.05
Description & Specifications

Specifications

Antigen CD75
Clone LN-1
Host Species Mouse
Species Reactivity Human
Applications Flow Cytometry
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.25μg)/test

This LN-1 monoclonal antibody reacts with human CD75, a cell surface carbohydrate determinant produced by alpha-2,6-sialytransferase. CD75 is expressed predominantly on mature B cells in peripheral blood and germinal centers, as well as several B cell lines (e.g., Raji, Ramos). CD75 is also expressed on non-immune tissues such as breast, prostate, colon, liver, and kidney. Reports demonstrate that CD75 is a ligand of CD22.

Studies have shown that, depending on fixation and tissue processing, the LN-1 antibody also recognizes astrocytes and microglia in the human brain.